COMH4211C2B1 Combi-Blocks, Inc.

1-[2-(Difluoromethoxy)benzyl]-2-methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-benzimidazole

  • MDL number MFCD31579552
  • Cas Number : 1515924-46-5
  • Molecular Weight : 414.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8557

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8557
Availability details for
Quantity
  • Availability
  • Add to Cart