COMH4211C2BF Combi-Blocks, Inc.

4-Chloro-2-[(cyclopropylmethyl)aminocarbonyl]pyridine

  • MDL number MFCD11177873
  • Cas Number : 417723-53-6
  • Molecular Weight : 210.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8581

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8581
Availability details for
Quantity
  • Availability
  • Add to Cart