COMH4211C2C0 Combi-Blocks, Inc.

5-Bromo-2-(2,2,2-trifluoroethoxy)benzaldehyde

  • MDL number MFCD11184432
  • Cas Number : 923281-72-5
  • Molecular Weight : 283.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8582

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8582
Availability details for
Quantity
  • Availability
  • Add to Cart