COMH4211C2C7 Combi-Blocks, Inc.

4-(2-Hydroxy-ethyl)-3,3-dimethyl-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31579391
  • Cas Number : 958026-99-8
  • Molecular Weight : 257.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8611

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8611
Availability details for
Quantity
  • Availability
  • Add to Cart