COMH4211C2EC Combi-Blocks, Inc.

4-(2,2-Difluoroethoxy)-3-methylbenzoic acid

  • MDL number MFCD24230951
  • Cas Number : 1373864-75-5
  • Molecular Weight : 216.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8626

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8626
Availability details for
Quantity
  • Availability
  • Add to Cart