COMH4211C2F5 Combi-Blocks, Inc.

1H-Indazole,5-bromo-4-(phenylmethoxy)-

  • MDL number MFCD13177086
  • Cas Number : 850363-66-5
  • Molecular Weight : 303.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8639

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8639
Availability details for
Quantity
  • Availability
  • Add to Cart