COMH4211C2FC Combi-Blocks, Inc.

1-(2,5-Dioxopyrrolidin-1-yl) 8-methyl octanedioate

  • MDL number MFCD29041539
  • Cas Number : 1609637-03-7
  • Molecular Weight : 285.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8654

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8654
Availability details for
Quantity
  • Availability
  • Add to Cart