COMH4211C307 Combi-Blocks, Inc.

3-(Cyclopropylmethoxy)-2-methylbenzoic acid

  • MDL number MFCD24311735
  • Cas Number : 1369797-94-3
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8669

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8669
Availability details for
Quantity
  • Availability
  • Add to Cart