COMH4211C30A Combi-Blocks, Inc.

2-(2-(Allyloxy)ethoxy)ethyl 4-methylbenzenesulfonate

  • MDL number MFCD13659293
  • Cas Number : 84183-96-0
  • Molecular Weight : 300.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8675

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8675
Availability details for
Quantity
  • Availability
  • Add to Cart