COMH4211C329 Combi-Blocks, Inc.

(E)-N-(2-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)but-2-enamide

  • MDL number MFCD31583235
  • Cas Number : 1912446-92-4
  • Molecular Weight : 301.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8753

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8753
Availability details for
Quantity
  • Availability
  • Add to Cart