COMH4211C32C Combi-Blocks, Inc.

6-Cyclopropanesulfonylamino-hexanoic acid

  • MDL number MFCD20352191
  • Cas Number : 1338943-82-0
  • Molecular Weight : 235.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8762

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8762
Availability details for
Quantity
  • Availability
  • Add to Cart