COMH4211C32F Combi-Blocks, Inc.

4-Bromo-n-cyclobutyl-3,5-difluoroaniline

  • MDL number MFCD31613861
  • Cas Number : 2015528-01-3
  • Molecular Weight : 262.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8773

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8773
Availability details for
Quantity
  • Availability
  • Add to Cart