COMH4211C357 Combi-Blocks, Inc.

2-(2-Bromoethoxy)-4-fluoro-1-(trifluoromethyl)benzene

  • MDL number MFCD31613704
  • Cas Number : 1953183-85-1
  • Molecular Weight : 287.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8805

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8805
Availability details for
Quantity
  • Availability
  • Add to Cart