COMH4211C371 Combi-Blocks, Inc.

(Cyclopropylmethyl)[1-(2,4-dichlorophenyl)ethyl]amine

  • MDL number MFCD11140373
  • Cas Number : 1019612-07-7
  • Molecular Weight : 244.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8879

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8879
Availability details for
Quantity
  • Availability
  • Add to Cart