COMH4211C387 Combi-Blocks, Inc.

3-Chloro-n-(cyclopropylmethyl)aniline

  • MDL number MFCD08544247
  • Cas Number : 939757-86-5
  • Molecular Weight : 181.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8923

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8923
Availability details for
Quantity
  • Availability
  • Add to Cart