COMH4211C391 Combi-Blocks, Inc.

2-[2-(Tetrahydro-pyran-2-yloxy)-ethoxy]-thiazole-4-carboxylic acid

  • MDL number MFCD31614052
  • Cas Number : 1448673-87-7
  • Molecular Weight : 273.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8947

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8947
Availability details for
Quantity
  • Availability
  • Add to Cart