COMH4211C395 Combi-Blocks, Inc.

4-Phenyl-1-[1,2,4]triazol-1-yl-butan-2-one

  • MDL number MFCD30145505
  • Cas Number : 1092851-29-0
  • Molecular Weight : 215.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8956

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8956
Availability details for
Quantity
  • Availability
  • Add to Cart