COMH4211C397 Combi-Blocks, Inc.

2-Chloro-3-(difluoromethoxy)benzaldehyde

  • MDL number MFCD27933739
  • Cas Number : 937069-63-1
  • Molecular Weight : 206.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8966

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8966
Availability details for
Quantity
  • Availability
  • Add to Cart