COMH4211C398 Combi-Blocks, Inc.

4-Methyl-3-(2-methylpropoxy)aniline

  • MDL number MFCD11934859
  • Cas Number : 1154268-71-9
  • Molecular Weight : 179.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8969

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8969
Availability details for
Quantity
  • Availability
  • Add to Cart