COMH4211C3B4 Combi-Blocks, Inc.

3-[(3,5-Difluorobenzyl)oxy]azetidine

  • MDL number MFCD11874828
  • Cas Number : 1121595-11-6
  • Molecular Weight : 199.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8984

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8984
Availability details for
Quantity
  • Availability
  • Add to Cart