COMH4211C3BB Combi-Blocks, Inc.

(4-Acetyl-phenoxy)-acetic acid tert-butyl ester

  • MDL number MFCD31618882
  • Cas Number : 165947-76-2
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9001

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9001
Availability details for
Quantity
  • Availability
  • Add to Cart