COMH4211C3BC Combi-Blocks, Inc.

4-Bromo-3-(difluoromethoxy)benzoic acid

  • MDL number MFCD28401976
  • Cas Number : 1352719-48-2
  • Molecular Weight : 267.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9002

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9002
Availability details for
Quantity
  • Availability
  • Add to Cart