COMH4211C3BD Combi-Blocks, Inc.

N-[4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]benzenesulfonamide

  • MDL number MFCD22494428
  • Cas Number : 914606-88-5
  • Molecular Weight : 359.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9003

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9003
Availability details for
Quantity
  • Availability
  • Add to Cart