COMH4211C3BE Combi-Blocks, Inc.

4-(Diethylcarbamoyl)-3-fluoro-benzoic acid

  • MDL number MFCD31583057
  • Cas Number : 1400643-60-8
  • Molecular Weight : 239.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9004

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9004
Availability details for
Quantity
  • Availability
  • Add to Cart