COMH4211C3C6 Combi-Blocks, Inc.

5-Amino-n-cyclopropylmethyl-4-fluoro-2-methyl-benzamide

  • MDL number MFCD31579297
  • Cas Number : 1852024-01-1
  • Molecular Weight : 222.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9022

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9022
Availability details for
Quantity
  • Availability
  • Add to Cart