COMH4211C3C7 Combi-Blocks, Inc.

1-[3-(2,2-Difluoro-ethoxy)-benzyl]-piperazine

  • MDL number MFCD31561469
  • Cas Number : 2270906-65-3
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9023

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9023
Availability details for
Quantity
  • Availability
  • Add to Cart