COMH4211C3C9 Combi-Blocks, Inc.

tert-Butyl 4-(4-amino-3-fluorophenoxy)piperidine-1-carboxylate

  • MDL number MFCD30988262
  • Cas Number : 1394808-79-7
  • Molecular Weight : 310.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9025

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9025
Availability details for
Quantity
  • Availability
  • Add to Cart