COMH4211C3CD Combi-Blocks, Inc.

1-(2,2-Difluoroethyl)piperidine-4-carbonitrile

  • MDL number MFCD20318364
  • Cas Number : 1343312-26-4
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9039

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9039
Availability details for
Quantity
  • Availability
  • Add to Cart