COMH4211C3CE Combi-Blocks, Inc.

(4-Ethoxy-pyrazol-1-yl)-acetic acid

  • MDL number MFCD31579319
  • Cas Number : 1862988-70-2
  • Molecular Weight : 170.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9040

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9040
Availability details for
Quantity
  • Availability
  • Add to Cart