COMH4211C3D3 Combi-Blocks, Inc.

1-(3-Bromo-benzyl)-4-ethoxy-1h-pyrazole

  • MDL number MFCD31561527
  • Cas Number : 1869783-70-9
  • Molecular Weight : 281.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9051

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9051
Availability details for
Quantity
  • Availability
  • Add to Cart