COMH4211C3D5 Combi-Blocks, Inc.

(3-Methoxy-propyl)-3-[4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenyl]-propyl-carbamic acid tert-butyl ester

  • MDL number MFCD31583157
  • Cas Number : 1318792-82-3
  • Molecular Weight : 433.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9054

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9054
Availability details for
Quantity
  • Availability
  • Add to Cart