COMH4211C3D6 Combi-Blocks, Inc.

Chloromethyl (2r)-2-[(tert-butoxy)carbonyl]aminobutanoate

  • MDL number MFCD30732394
  • Cas Number : 2137034-08-1
  • Molecular Weight : 251.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9057

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9057
Availability details for
Quantity
  • Availability
  • Add to Cart