COMH4211C3D9 Combi-Blocks, Inc.

3-Bromo-n-cyclopropyl-5-trifluoromethyl-benzamide

  • MDL number MFCD31619009
  • Cas Number : 2270908-68-2
  • Molecular Weight : 308.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9065

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9065
Availability details for
Quantity
  • Availability
  • Add to Cart