COMH4211C3DE Combi-Blocks, Inc.

1-[4-(2,2-Difluoro-ethoxy)-phenyl]-propan-2-one

  • MDL number MFCD24220037
  • Cas Number : 1553316-34-9
  • Molecular Weight : 214.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9074

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9074
Availability details for
Quantity
  • Availability
  • Add to Cart