COMH4211C3DF Combi-Blocks, Inc.

(4-Bromo-3-difluoromethoxy-phenyl)-methanol

  • MDL number MFCD23711898
  • Cas Number : 1331943-89-5
  • Molecular Weight : 253.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9075

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9075
Availability details for
Quantity
  • Availability
  • Add to Cart