COMH4211C3E0 Combi-Blocks, Inc.

3-Fluoro-4-(propane-2-sulfonyl)aniline

  • MDL number MFCD09909522
  • Cas Number : 1048918-84-8
  • Molecular Weight : 217.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9076

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9076
Availability details for
Quantity
  • Availability
  • Add to Cart