COMH4211C3E6 Combi-Blocks, Inc.

[2-Methyl-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)phenyl]-morpholin-4-yl-methanone

  • MDL number MFCD26398862
  • Cas Number : 1013643-15-6
  • Molecular Weight : 331.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9085

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9085
Availability details for
Quantity
  • Availability
  • Add to Cart