COMH4211C3F0 Combi-Blocks, Inc.

3-Bromo-4-(4-methoxy-benzyloxy)-benzonitrile

  • MDL number MFCD25801513
  • Cas Number : 1548389-21-4
  • Molecular Weight : 318.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9104

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9104
Availability details for
Quantity
  • Availability
  • Add to Cart