COMH4211C3F1 Combi-Blocks, Inc.

4-(4-Fluorobenzyloxy)-1h-pyridin-2-one

  • MDL number MFCD24549418
  • Cas Number : 586373-38-8
  • Molecular Weight : 219.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9108

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9108
Availability details for
Quantity
  • Availability
  • Add to Cart