COMH4211C3FC Combi-Blocks, Inc.

[(2,4-Difluorophenyl)methyl](methoxy)amine

  • MDL number MFCD21116563
  • Cas Number : 543731-20-0
  • Molecular Weight : 173.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9139

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9139
Availability details for
Quantity
  • Availability
  • Add to Cart