COMH4211C419 Combi-Blocks, Inc.

3-(1-Ethanesulfonyl-piperidin-4-yl)-propionic acid

  • MDL number MFCD16723176
  • Cas Number : 1250510-43-0
  • Molecular Weight : 249.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9140

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9140
Availability details for
Quantity
  • Availability
  • Add to Cart