COMH4211C41E Combi-Blocks, Inc.

2-(2-Isobutoxyphenyl)ethanol

  • MDL number MFCD14582729
  • Cas Number : 1378825-15-0
  • Molecular Weight : 194.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9149

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9149
Availability details for
Quantity
  • Availability
  • Add to Cart