COMH4211C41F Combi-Blocks, Inc.

1-Bromo-3-(cyclopropylmethoxy)-5-fluorobenzene

  • MDL number MFCD23500338
  • Cas Number : 1369948-31-1
  • Molecular Weight : 245.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9150

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9150
Availability details for
Quantity
  • Availability
  • Add to Cart