COMH4211C423 Combi-Blocks, Inc.

Methyl 6-(benzyloxy)-3-oxohexanoate

  • MDL number MFCD31583043
  • Cas Number : 299171-56-5
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9159

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9159
Availability details for
Quantity
  • Availability
  • Add to Cart