COMH4211C42F Combi-Blocks, Inc.

2-(4-Nitro-phenoxycarbonyloxy)-propionic acid ethyl ester

  • MDL number MFCD31583359
  • Cas Number : 2270913-05-6
  • Molecular Weight : 283.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9181

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9181
Availability details for
Quantity
  • Availability
  • Add to Cart