COMH4211C431 Combi-Blocks, Inc.

4-Bromo-n-(2-hydroxyethyl)-3-(2,2,2-trifluoroethoxy)benzamide

  • MDL number MFCD31539509
  • Cas Number : 1309682-55-0
  • Molecular Weight : 342.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9183

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9183
Availability details for
Quantity
  • Availability
  • Add to Cart