COMH4211C432 Combi-Blocks, Inc.

9-Iodo-1-nonanol

  • MDL number MFCD30729433
  • Cas Number : 76334-30-0
  • Molecular Weight : 270.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9185

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9185
Availability details for
Quantity
  • Availability
  • Add to Cart