COMH4211C437 Combi-Blocks, Inc.

Benzoic acid, 3-(methoxymethoxy)-, 2,5-dioxo-1-pyrrolidinyl ester

  • MDL number MFCD31381654
  • Cas Number : 1560874-18-1
  • Molecular Weight : 279.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9196

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9196
Availability details for
Quantity
  • Availability
  • Add to Cart