COMH4211C43C Combi-Blocks, Inc.

N-(5-Amino-2-chlorophenyl)-2-(morpholin-4-yl)acetamide

  • MDL number MFCD09673360
  • Cas Number : 928712-73-6
  • Molecular Weight : 269.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9208

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9208
Availability details for
Quantity
  • Availability
  • Add to Cart