COMH4211C43F Combi-Blocks, Inc.

1-Methanesulfonyloxymethyl-cyclopropanecarboxylic acid ethyl ester

  • MDL number MFCD31381699
  • Cas Number : 633335-73-6
  • Molecular Weight : 222.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9213

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9213
Availability details for
Quantity
  • Availability
  • Add to Cart